Learn More
Methyl phenyl sulfoxide, 98+%
Product Code. 11427247
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11427247
Brand: Thermo Scientific Alfa Aesar A15009.14
Request Bulk or Custom Format
CAS: 1193-82-4 | C7H8OS | 140.2 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00002088
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1
Specifications
Melting Point 26°C to 32°C
Boiling Point 93°C to 94°C (0.3 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.5775
Quantity 25 g
Beilstein 1904976
Sensitivity Hygroscopic
Formula Weight 140.2
Percent Purity ≥98%
Chemical Name or Material Methyl phenyl sulfoxide