Learn More
Methyl Pentadecafluoroheptyl Ketone, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Pentadecafluoroheptyl Ketone |
| CAS | 754-85-8 |
| Molecular Formula | C9H3OF15 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C9H3F15O/c1-2(25)3(10,11)4(12,13)5(14,15)6(16,17)7(18,19)8(20,21)9(22,23)24/h1H3 |
| SMILES | CC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-pentadecafluorononan-2-one |
| Molecular Weight (g/mol) | 412.1 |
| Formula Weight | 411.9944 |