Learn More
Methyl Octadecanoate-d35, CDN
Specifications
Specifications
| Chemical Name or Material | Octadecanoic Acid Methyl Ester-D35 (octadecanoic-D35) |
| CAS | 29823-25-4 |
| Molecular Formula | C19 D35 H3 O2 |
| Quantity | 0.1 g |
| Synonym | Octadecanoic-d35 acid, methyl ester (9 CI), Stearic-d35 acid, methyl ester (8 CI), Octadecanoic-d35 acid methyl ester, Stearic-d35 acid methyl ester, Methyl Stearate-d35, Methyl octadecanoate-d35, Octadecanoic acid methyl ester-D35 (octadecanoic-D35) |
| InChI Formula | InChI=1 S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC |
| IUPAC Name | methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,18-pentatriacontadeuteriooctadecanoate |
| Molecular Weight (g/mol) | 333.7195 |
| Formula Weight | 333.5069 g/mol |
| Show More |