missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Nonanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Nonanoic acid methyl ester |
| CAS | 1731-84-6 |
| Molecular Formula | C10 H20 O2 |
| Quantity | 1 g |
| Synonym | Nonanoic acid-methyl ester, Nonanoic acid, methyl ester (6CI, 8CI, 9CI, ACI), Methyl n-nonanoate, Methyl nonanoate, Methyl nonylate, Methyl pelargonate, Pelargonic acid methyl ester |
| InChI Formula | InChI=1S/C10H20O2/c1-3-4-5-6-7-8-9-10(11)12-2/h3-9H2,1-2H3 |
| SMILES | CCCCCCCCC(=O)OC |
| IUPAC Name | methyl nonanoate |
| Molecular Weight (g/mol) | 172.26 |
| Formula Weight | 172.1463 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?