missing translation for 'onlineSavingsMsg'
Learn More

Methyl Nonanoate, TRC

Product Code. 30468922
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468922 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468922 Supplier Toronto Research Chemicals Supplier No. TRCM2255855G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nonanoic acid methyl ester
CAS 1731-84-6
Molecular Formula C10 H20 O2
Quantity 5 g
Synonym Nonanoic acid-methyl ester, Nonanoic acid, methyl ester (6CI, 8CI, 9CI, ACI), Methyl n-nonanoate, Methyl nonanoate, Methyl nonylate, Methyl pelargonate, Pelargonic acid methyl ester
InChI Formula InChI=1S/C10H20O2/c1-3-4-5-6-7-8-9-10(11)12-2/h3-9H2,1-2H3
SMILES CCCCCCCCC(=O)OC
IUPAC Name methyl nonanoate
Molecular Weight (g/mol) 172.26
Formula Weight 172.1463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.