missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Jasmonate-d5 (2,4,4-d3; acetyl-2,2-d2) (mixture of diastereomers), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Methyl Jasmonate-d5 (2,4,4-d3; acetyl-2,2-d2) (mixture of diastereomers) |
| Molecular Formula | C13 D5 H15 O3 |
| Quantity | 0.01 g |
| Synonym | 3-Oxo-2-(2 Z)-2-penten-1-ylcyclopentaneacetic-α,α,2,4,4-d5 acid methyl ester, Methyl Jasmonate-d5 (2,4,4-d3, acetyl-2,2-d2) (mixture of diastereomers), Jasmonic acid-2,2,5,5,7-d5 methyl ester (mixture of isomers), Jasmonic Acid Methyl Ester-D5 (cyclopentaneacetic-α,α,2,4,4-D5) (mixture of isomers) |
| InChI Formula | InChI=1 S/C13H20O3/c1-3-4-5-6-11-10(7-8-12(11)14)9-13(15)16-2/h4-5,10-11 H,3,6-9H2,1-2H3/b5-4-/i8D2,9D2,11 D |
| SMILES | [2 H]C([2 H])(C1CC([2 H])([2 H])C(=O)C1([2 H])C\C=C/CC)C(=O)OC |
| IUPAC Name | methyl 2,2-dideuterio-2-[2,4,4-trideuterio-3-oxo-2-[(Z)-pent-2-enyl]cyclopentyl]acetate |
| Molecular Weight (g/mol) | 229.3269 |
| Formula Weight | 229.1726 g/mol |
| Percent Purity | 96 atom % D, min 96% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction