missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isodehydracetate, TRC

Product Code. 30470961
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470961 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470961 Supplier Toronto Research Chemicals Supplier No. TRCM31410025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Isodehydracetate
CAS 41264-06-6
Molecular Formula C9 H10 O4
Quantity 25 g
Synonym 2H-Pyran-5-carboxylic acid','4,6-dimethyl-2-oxo-','methyl ester, Methyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate, 2,4-Dimethyl-6-oxo-6H-pyran-3-carboxylic acid methyl ester, Isodehydroacetic acid methyl ester, Methyl 4,6-dimethyl-2-pyrone-5-carboxylate, Methyl isodehydracetate, Methyl isodehydroacetate
InChI Formula InChI=1S/C9H10O4/c1-5-4-7(10)13-6(2)8(5)9(11)12-3/h4H,1-3H3
SMILES COC(=O)C1=C(C)OC(=O)C=C1C
IUPAC Name methyl 2,4-dimethyl-6-oxopyran-3-carboxylate
Molecular Weight (g/mol) 182.17
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.