Learn More
Methyl Hexadecanoate-d31, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.10g
0.50g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic Acid Methyl Ester-D31 (hexadecanoic-D31) |
| CAS | 29848-79-1 |
| Molecular Formula | C17 D31 H3 O2 |
| Quantity | 0.1 g |
| Synonym | Methyl Palmitate-d31, Hexadecanoic-d31 acid, methyl ester (9 CI), Palmitic-d31 acid, methyl ester (8 CI), Methyl perdeuteriohexadecanoate, Methyl Hexadecanoate-d31, Hexadecanoic-d31 acid methyl ester, Palmitic-d31 acid methyl ester, Hexadecanoic acid methyl ester-D31 (hexadecanoic-D31) |
| InChI Formula | InChI=1 S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC |
| IUPAC Name | methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-hentriacontadeuteriohexadecanoate |
| Molecular Weight (g/mol) | 301.6417 |
| Formula Weight | 301.4505 g/mol |
| Show More |