missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Heptanoate-6,6,7,7,7-d5, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Methyl Heptanoate-6,6,7,7,7-D5 |
| Molecular Formula | C8 D5 H11 O2 |
| Quantity | 0.1 g |
| Synonym | Methyl heptanoate-6,6,7,7,7-d5, Heptanoic-6,6,7,7,7-d5 acid methyl ester, Heptanoic acid methyl ester-d5, Methyl heptanoate-6,6,7,7,7-D5, Methyl heptanoate-d5 |
| InChI Formula | InChI=1 S/C8H16O2/c1-3-4-5-6-7-8(9)10-2/h3-7H2,1-2H3/i1D3,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCC(=O)OC |
| IUPAC Name | methyl 6,6,7,7,7-pentadeuterioheptanoate |
| Molecular Weight (g/mol) | 149.2423 |
| Formula Weight | 149.1464 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction