Learn More
Methyl Ethanesulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Ethanesulfonate |
| CAS | 1912-28-3 |
| Molecular Formula | C3H8O3S |
| Quantity | 1 g |
| Synonym | Methyl Ethylsulfonate, Ethanesulfonic Acid Methyl Ester |
| InChI Formula | InChI=1S/C3H8O3S/c1-3-7(4,5)6-2/h3H2,1-2H3 |
| SMILES | COS(CC)(=O)=O |
| IUPAC Name | methyl ethanesulfonate |
| Molecular Weight (g/mol) | 124.16 |
| Formula Weight | 124.0194 |