Learn More
Methyl-d2 Alcohol, CDN
Product Code. 30316968
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316968 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316968 Supplier CDN Supplier No. CDND07581G
 Restricted Item
Estimated Shipment: 09-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methanol-1,1-D2
CAS 28563-35-1
Molecular Formula C D2 H2 O
Quantity 1 g
Synonym Methan-d2-ol (8 CI, 9 CI), Dideuteriomethanol, Methanol-d2 (CHD2OH), Methyl-d2 alcohol, Methanol-1,1-D2
InChI Formula InChI=1 S/CH4O/c1-2/h2H,1H3/i1D2
SMILES [2 H]C([2 H])O
IUPAC Name dideuteriomethanol
Molecular Weight (g/mol) 34.0542
Formula Weight 34.0388 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity