Learn More
Methyl benzo[b]thiophene-2-carboxylate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 22913-24-2 |
| Molecular Formula | C10H8O2S |
| MDL Number | MFCD00067791 |
| Quantity | 100 g |
| SMILES | O=C(C1=CC2=CC=CC=C2S1)OC |
| Molecular Weight (g/mol) | 192.23 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl benzo[b]thiophene-2-carboxylate |