missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-(trifluoromethyl)nicotinate, 97%, Thermo Scientific™

Product Code. 15486707
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486707 250 mg 250mg
15466707 1 g 1g
15476707 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15486707 Supplier Thermo Scientific Alfa Aesar Supplier No. H33836.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 6-(trifluoromethyl)nicotinate is used as a pharmaceutical intermediate.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221313-10-6
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.136
MDL Number MFCD03001346
InChI Key NEATXKNYTRXHQL-UHFFFAOYSA-N
PubChem CID 2775647
IUPAC Name methyl 6-(trifluoromethyl)pyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)C(F)(F)F

Specifications

Melting Point 54°C to 56°C
Quantity 250 mg
Synonym methyl 6-trifluoromethyl nicotinate, methyl 6-trifluoromethyl pyridine-3-carboxylate, methyl6-trifluoromethyl nicotinate, 3-pyridinecarboxylic acid, 6-trifluoromethyl-, methyl ester, methyl 6-trifluoromethylnicotinate, methyl 2-trifluoromethyl pyridine-5-carboxylate, methyl 2-trifluoromethyl-5-pyridinecarboxylate, 6-trifluoromethylnicotinic acid methyl ester, pubchem18762, methyltrifluoromethylnicotinate
Solubility Information Soluble in acetone. Insoluble in water.
Formula Weight 205.14
Percent Purity 97%
Chemical Name or Material Methyl 6-(trifluoromethyl)nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.