Learn More
methyl 5-(hydroxymethyl)furan-2-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | methyl 5-(hydroxymethyl)furan-2-carboxylate |
| CAS | 36802-01-4 |
| Molecular Formula | C7H8O4 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C7H8O4/c1-10-7(9)6-3-2-5(4-8)11-6/h2-3,8H,4H2,1H3 |
| SMILES | COC(=O)c1oc(CO)cc1 |
| IUPAC Name | methyl 5-(hydroxymethyl)furan-2-carboxylate |
| Molecular Weight (g/mol) | 156.14 |
| Formula Weight | 156.0423 |