Learn More
Methyl 5-bromonicotinate, 97%, Thermo Scientific Chemicals
Product Code. 11449946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449946 5 g 5g
11439946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11449946 Supplier Thermo Scientific Alfa Aesar Supplier No. B20908.06
 Restricted Item
In Stock
Add to basket
CAS: 29681-44-5 | Molecular Formula: C7H6BrNO2 | Molecular Weight: 216.034 g/mol
A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29681-44-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00173668
InChI Key AAJZXPWBILCHAW-UHFFFAOYSA-N
Synonym methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
PubChem CID 699336
IUPAC Name methyl 5-bromopyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1)Br
Specifications
Melting Point 96°C to 98°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 216.04
Percent Purity 97%
Chemical Name or Material Methyl 5-bromonicotinate