Learn More
Methyl 4-oxo-4-phenylbutanoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 25333-24-8 |
| Molecular Formula | C11H12O3 |
| MDL Number | MFCD00014906 |
| Quantity | 25 g |
| SMILES | O=C(OC)CCC(C1=CC=CC=C1)=O |
| Molecular Weight (g/mol) | 192.21 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 4-oxo-4-phenylbutanoate |