missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Deoxy-4-fluoro-Alpha-D-glucose, TRC

Product Code. 30179493
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179493 5 mg 5mg
30179001 25 mg 25mg
30179581 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179493 Supplier Toronto Research Chemicals Supplier No. TRCM3052755MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56926-53-5
Molecular Formula C7 H13 F O5
Molecular Weight (g/mol) 196.17
Synonym Methyl 4-Deoxy-4-Fluoro-α-D-Glucose
IUPAC Name (2S,3R,4R,5S,6R)-5-fluoro-6-(hydroxymethyl)-2-methoxyoxane-3,4-diol
SMILES CO[C@H]1O[C@H](CO)[C@@H](F)[C@H](O)[C@H]1O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C7H13FO5/c1-12-7-6(11)5(10)4(8)3(2-9)13-7/h3-7,9-11H,2H2,1H3/t3-,4-,5+,6-,7+/m1/s1
Formula Weight 196.0747
Chemical Name or Material Methyl 4-Deoxy-4-fluoro-a-D-glucose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.