Learn More
Methyl 4-Amino-2-methoxy-5-thiocyanobenzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 4-amino-2-methoxy-5-thiocyanobenzoate |
| CAS | 59168-56-8 |
| Molecular Formula | C10 H10 N2 O3 S |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C10H10N2O3S/c1-14-8-4-7(12)9(16-5-11)3-6(8)10(13)15-2/h3-4H,12H2,1-2H3 |
| SMILES | COC(=O)c1cc(SC#N)c(N)cc1OC |
| IUPAC Name | methyl 4-amino-2-methoxy-5-thiocyanatobenzoate |
| Molecular Weight (g/mol) | 238.26 |
| Formula Weight | 238.0412 |