Learn More
Methyl 4-Amino-2,3,5,6-tetrafluorobenzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 4-Amino-2,3,5,6-tetrafluorobenzoate |
| CAS | 715-37-7 |
| Molecular Formula | C8H5NO2F4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C8H5F4NO2/c1-15-8(14)2-3(9)5(11)7(13)6(12)4(2)10/h13H2,1H3 |
| SMILES | COC(=O)c1c(F)c(F)c(N)c(F)c1F |
| IUPAC Name | methyl 4-amino-2,3,5,6-tetrafluorobenzoate |
| Molecular Weight (g/mol) | 223.12 |
| Formula Weight | 223.0256 |