Learn More
Methyl 3alpha,7alpha-Diacetoxy-12-oxo-5beta-cholan-24-oate, TRC
Product Code. 30482767
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482767 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30482767 Supplier Toronto Research Chemicals Supplier No. TRCM303505100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 3alpha,7alpha-Diacetoxy-12-oxo-5beta-cholan-24-oate
CAS 28535-81-1
Molecular Formula C29 H44 O7
Quantity 100 mg
Synonym Cholan-24-oic acid','3,7-bis(acetyloxy)-12-oxo-','methyl ester','(3α,5β,7α)-, 5β-Cholan-24-oic acid','3α,7α-dihydroxy-12-oxo-','methyl ester','diacetate (8CI), 5β-Cholanic acid','3α,7α-dihydroxy-12-oxo-','methyl ester','diacetate (6CI,7CI), 3α,7α-Diacetoxy-12-oxo-5β-cholan-24-oic acid methyl ester, Methyl 3α,7α-diacetoxy-12-oxo-5β-cholan-24-oate, Methyl 3α,7α-diacetoxy-12-oxo-5β-cholanate
InChI Formula InChI=1S/C29H44O7/c1-16(7-10-26(33)34-6)21-8-9-22-27-23(15-25(32)29(21,22)5)28(4)12-11-20(35-17(2)30)13-19(28)14-24(27)36-18(3)31/h16,19-24,27H,7-15H2,1-6H3/t16-,19+,20-,21-,22+,23+,24-,27+,28+,29-/m1/s1
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@@H](C[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3CC(=O)[C@]12C)OC(=O)C)OC(=O)C
IUPAC Name methyl (4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-diacetyloxy-10,13-dimethyl-12-oxo-1,2,3,4,5,6,7,8,9,11,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 504.66
Formula Weight 504.3087