Learn More
Methyl 3-Phenyloxirane-2-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 3-Phenyloxirane-2-carboxylate |
| CAS | 37161-74-3 |
| Molecular Formula | C10H10O3 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C10H10O3/c1-12-10(11)9-8(13-9)7-5-3-2-4-6-7/h2-6,8-9H,1H3 |
| SMILES | COC(=O)C1OC1c2ccccc2 |
| IUPAC Name | methyl 3-phenyloxirane-2-carboxylate |
| Molecular Weight (g/mol) | 178.18 |
| Formula Weight | 178.063 |