Learn More
Methyl 3-oxo-pentanoate, 99+%
Product Code. 10752384 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
Product Code. 10752384
Brand: Thermo Scientific Acros 243161000
Request Bulk or Custom Format
CAS: 30414-53-0 | C6H10O3 | 130.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30414-53-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00011705
InChI Key XJMIXEAZMCTAGH-UHFFFAOYSA-N
Synonym methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
PubChem CID 121699
IUPAC Name methyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OC
Specifications
Melting Point -35°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 95°C to 97°C (48.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CH2COCH2COOCH3
Refractive Index 1.4220 to 1.424
Quantity 100 mL
Fieser 10,415; 01,1022; 04,437; 05,589; 07,324; 11,472; 12,126; 16,303
Specific Gravity 1.03
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with most common organic solvents
Formula Weight 130.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Methyl 3-oxo-pentanoate, 99+%