Learn More
Methyl 3-oxo-4-phenylbutanoate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 37779-49-0 |
| Molecular Formula | C11H12O3 |
| MDL Number | MFCD03424746 |
| Quantity | 10 g |
| SMILES | COC(=O)CC(=O)CC1=CC=CC=C1 |
| Molecular Weight (g/mol) | 192.21 |
| Percent Purity | 95% |
| Chemical Name or Material | Methyl 3-oxo-4-phenylbutanoate |