missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chloro-4-methoxybenzoate, 98%, Thermo Scientific™

Product Code. p-7021141
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11490983

Brand: Thermo Scientific Alfa Aesar A13356.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37908-98-8
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017188
InChI Key PINQDVFQCCFACD-UHFFFAOYSA-N
Synonym methyl 3-chloro-p-anisate, 3-chloro-4-methoxybenzoic acid methyl ester, methyl3-chloro-4-methoxybenzoate, benzoic acid, 3-chloro-4-methoxy-, methyl ester, methyl-3-chlor-anisat, acmc-1aehy, rarechem al bf 0700, methyl-3-chloro-4-methoxybenzoate, 2-chloro-4-methoxycarbonyl anisole, 3-cl-4-ch3o-c6h3-cooch3
PubChem CID 123469
IUPAC Name methyl 3-chloro-4-methoxybenzoate
SMILES COC1=C(C=C(C=C1)C(=O)OC)Cl

Specifications

Melting Point 93°C to 97°C
Quantity 10 g
Beilstein 2365816
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material Methyl 3-chloro-4-methoxybenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.