Learn More
Methyl 3-bromo-5-hydroxybenzoate, 97%, Thermo Scientific Chemicals
Product Code. p-7034076
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15478177
Brand: Thermo Scientific Alfa Aesar H51724.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 192810-12-1
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD10566800
InChI Key KRTBWIIGEJIUSA-UHFFFAOYSA-N
Synonym 3-bromo-5-hydroxybenzoic acid methyl ester, acmc-209f1w, methyl3-bromo-5-hydroxybenzoate, methyl 3-bromo-5-hydroxylbenzoate, 3-bromo-5-hydroxy-benzoic acid methyl ester, benzoic acid,3-bromo-5-hydroxy-,methyl ester, benzoic acid, 3-bromo-5-hydroxy-, methyl ester
PubChem CID 53216607
IUPAC Name methyl 3-bromo-5-hydroxybenzoate
SMILES COC(=O)C1=CC(=CC(=C1)Br)O
Specifications
Melting Point 130°C to 135°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 231.05
Percent Purity 97%
Chemical Name or Material Methyl 3-bromo-5-hydroxybenzoate