Learn More
Methyl 3,4-difluorobenzoate, 97%, Thermo Scientific™
Product Code. 11488697
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11488697
Brand: Thermo Scientific Alfa Aesar A19866.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 369-25-5
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD00276597
InChI Key DWRVHDWKWKFSAI-UHFFFAOYSA-N
Synonym methyl-3,4-difluorobenzoate, methyl 3,4-difluorobenzenecarboxylate, benzoic acid, 3,4-difluoro-, methyl ester, methyl3,4-difluorobenzoate, pubchem14795, methyl 3,4 difluorobenzoate, acmc-1cs42, methyldifluorobenzenecarboxylate, rarechem al bf 0215, buttpark 89\07-66
PubChem CID 2775358
IUPAC Name methyl 3,4-difluorobenzoate
SMILES COC(=O)C1=CC=C(F)C(F)=C1
Specifications
Boiling Point 87°C to 89°C (15 mmHg)
Quantity 5 g
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material Methyl 3,4-difluorobenzoate