Learn More
Methyl 2-chloro-4-fluorobenzoate, 97%, Thermo Scientific™
Product Code. 11433907
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433907 5 g 5g
11423907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11433907 Supplier Thermo Scientific Alfa Aesar Supplier No. A18342.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85953-29-3
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00173937
InChI Key YZDLPZNWBRBZMZ-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzoic acid methyl ester, methyl 2-chloro-4-fluorobenzenecarboxylate, benzoic acid, 2-chloro-4-fluoro-, methyl ester, 2-chloro-4-fluoro-benzoic acid methyl ester, pubchem18760, rarechem al bf 0289, acmc-209q7x, ksc495c1p, methyl 2-chloro-4-fluorob..., methy 2-chloro-4-fluorobenzoate
PubChem CID 2763551
IUPAC Name methyl 2-chloro-4-fluorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)Cl
Specifications
Boiling Point 103°C to 105°C (15 mmHg)
Quantity 5 g
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material Methyl 2-chloro-4-fluorobenzoate