missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloro-3-methoxypropionate, TRC

Product Code. 30475174
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475174 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475174 Supplier Toronto Research Chemicals Supplier No. TRCM2971002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-chloro-3-methoxypropionate
CAS 36997-03-2
Molecular Formula C5 H9 Cl O3
Quantity 2.5 g
Synonym α-Chlor-β-methoxypropansaeure-methylester
InChI Formula InChI=1S/C5H9ClO3/c1-8-3-4(6)5(7)9-2/h4H,3H2,1-2H3
SMILES COCC(Cl)C(=O)OC
IUPAC Name methyl 2-chloro-3-methoxypropanoate
Molecular Weight (g/mol) 152.576
Formula Weight 152.024
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.