Learn More
Methyl 2-amino-4-chlorobenzoate, 98%
Product Code. 11397566
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11397566
Brand: Thermo Scientific Alfa Aesar L11125.18
Request Bulk or Custom Format
CAS: 5900-58-3 | C8H8ClNO2 | 185.607 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5900-58-3
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00017568
InChI Key YPSSCICDVDOEAI-UHFFFAOYSA-N
Synonym methyl 4-chloroanthranilate, 2-amino-4-chloro-benzoic acid methyl ester, methyl2-amino-4-chlorobenzoate, benzoic acid, 2-amino-4-chloro-, methyl ester, 2-amino-4-chlorobenzoic acid methyl ester, pubchem4643, methyl 4-chloro-anthranilate, methyl 2-amino4-chlorobenzoate, methyl-2-amino-4-chlorobenzoate
PubChem CID 80001
IUPAC Name methyl 2-amino-4-chlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Cl)N
Specifications
Melting Point 66°C to 68°C
Quantity 50 g
Beilstein 1072895
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4-chlorobenzoate