Learn More
Methyl 2-Acetamidoacrylate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Methyl 2-Acetamidoacrylate |
| CAS | 35356-70-8 |
| Molecular Formula | C6H9NO3 |
| Quantity | 5 g |
| Synonym | 2-(Acetylamino)-2-propenoic Acid Methyl Ester, 2-Acetamidoacrylic Acid Methyl Ester, 2-(Acetylamino)-2-propenoic Acid Methyl Ester, 2-Acetylaminoacrylic Acid Methyl Ester, Methyl 2-(Acetylamino)acrylate, Methyl 2-Acetylamino-2-propenoate, Methyl alpha-Acetamidoacrylate, N-(1-(Methoxycarbonyl)vinyl)acetamide, N-(2-Methoxy-1-methylene-2-oxoethyl)acetamide, N-Acetyldehydroalanine Methyl Ester, NSC 156878 |
| InChI Formula | InChI=1S/C6H9NO3/c1-4(6(9)10-3)7-5(2)8/h1H2,2-3H3,(H,7,8) |
| SMILES | COC(=O)C(=C)NC(=O)C |
| IUPAC Name | methyl 2-acetamidoprop-2-enoate |
| Molecular Weight (g/mol) | 143.14 |
| Formula Weight | 143.0582 |