Learn More
Methyl 2-acetamidoacrylate, 97%
Product Code. 11392407
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11392407
Brand: Thermo Scientific Alfa Aesar L10049.03
Request Bulk or Custom Format
CAS: 35356-70-8 | C6H9NO3 | 143.14 g/mol
Methyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35356-70-8
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
MDL Number MFCD00013394
InChI Key SMWNFFKPVLVOQQ-UHFFFAOYSA-N
Synonym methyl 2-acetamidoacrylate, 2-acetamidoacrylic acid methyl ester, n-acetyldehydroalanine methyl ester, 2-propenoic acid, 2-acetylamino-, methyl ester, methyl 2-acetylamino acrylate, 2-acetamidomethacrylate, acmc-209igf, methyl 2-acetaminoacrylate, methyl 2-acetylamino-acrylate, methyl 2-acetamido-2-propenate
PubChem CID 98644
IUPAC Name methyl 2-acetamidoprop-2-enoate
SMILES COC(=O)C(=C)NC(C)=O
Specifications
Melting Point 49°C to 51°C
Boiling Point 103°C to 104°C (8 mmHg)
Quantity 1 g
Beilstein 1812208
Formula Weight 143.14
Percent Purity 99%
Chemical Name or Material Methyl 2-acetamidoacrylate