Learn More
Methyl 2,3,5-Tri-O-p-chlorobenzoyl-Beta-D-ribofuranoside, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 2,3,5-Tri-O-p-chlorobenzoyl-beta-D-ribofuranoside |
| CAS | 29755-00-8 |
| Molecular Formula | C27 H21 Cl3 O8 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C27H21Cl3O8/c1-34-27-23(38-26(33)17-6-12-20(30)13-7-17)22(37-25(32)16-4-10-19(29)11-5-16)21(36-27)14-35-24(31)15-2-8-18(28)9-3-15/h2-13,21-23,27H,14H2,1H3/t21-,22-,23-,27-/m1/s1 |
| SMILES | CO[C@@H]1O[C@H](COC(=O)c2ccc(Cl)cc2)[C@@H](OC(=O)c3ccc(Cl)cc3)[C@H]1OC(=O)c4ccc(Cl)cc4 |
| IUPAC Name | [(2R,3R,4R,5R)-3,4-bis[(4-chlorobenzoyl)oxy]-5-methoxyoxolan-2-yl]methyl 4-chlorobenzoate |
| Molecular Weight (g/mol) | 579.81 |
| Formula Weight | 578.0302 |