Learn More
Methyl 1-Testosterone, TRC
Specifications
Specifications
| Chemical Name or Material | 17b-Hydroxy-17a-methyl-5a-androst-1-ene-3-one |
| CAS | 65-04-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20H30O2 |
| Quantity | 250 mg |
| Synonym | 17β-Hydroxy-17α-methyl-5α-androst-1-ene-3-one, Methyl 1-Testosterone, 5α-Androst-1-en-3-one, 17β-hydroxy-17-methyl- (6CI,7CI,8CI), 17-Methyl-1-testosterone, 17α-Methyl-17β-hydroxy-5α-androst-1-en-3-one, 17α-Methyl-17β-hydroxy-Δ1-5α-androsten-3-one, 17β-Hydroxy-17-methyl-5α-androst-1-en-3-one, 17β-Hydroxy-17α-methyl-5α-androst-1-en-3-one, 17β-Hydroxy-17α-methyl-5α-androstan-1-en-3-one, Methyl-1-testosterone, SC 11195 |
| InChI Formula | InChI=1S/C20H30O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,13,15-17,22H,4-5,7-8,10-12H2,1-3H3/t13-,15+,16-,17-,18-,19-,20-/m0/s1 |
| SMILES | C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (5S,8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13,17-trimethyl-5,6,7,8,9,11,12,14,15,16-decahydro-4H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 302.4 |
| Show More |