Learn More
Methyl 1-Naphthaleneacetate, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Naphthyl acetic acid methyl ester |
| CAS | 2876-78-0 |
| Molecular Formula | C13 H12 O2 |
| Quantity | 2.5 g |
| Synonym | 1-Naphthyl acetic acid-methyl ester, 1-Naphthaleneacetic acid, methyl ester (6CI, 7CI, 8CI, 9CI, ACI), Hormonit, Kartofin, M 1, M 1 (growth regulator), Methyl 1-naphthaleneacetate, Methyl 1-naphthalenylacetate, Methyl 1-naphthylacetate, Methyl α-naphthaleneacetate, Methyl α-naphthylacetate, NSC 122030, α-Naphthaleneacetic acid methyl ester, α-Naphthylacetic acid methyl ester |
| InChI Formula | InChI=1S/C13H12O2/c1-15-13(14)9-11-7-4-6-10-5-2-3-8-12(10)11/h2-8H,9H2,1H3 |
| SMILES | COC(=O)Cc1cccc2ccccc12 |
| IUPAC Name | methyl 2-naphthalen-1-ylacetate |
| Molecular Weight (g/mol) | 200.23 |
| Formula Weight | 200.0837 |