Learn More
Methyl 1-(2,4-Difluorophenyl)-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 1-(2,4-Difluorophenyl)-3-(thiophen-2-yl)-1H-pyrazole-5-carboxylate |
| CAS | 1202028-96-3 |
| Molecular Formula | C15 H10 F2 N2 O2 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C15H10F2N2O2S/c1-21-15(20)13-8-11(14-3-2-6-22-14)18-19(13)12-5-4-9(16)7-10(12)17/h2-8H,1H3 |
| SMILES | COC(=O)c1cc(nn1c2ccc(F)cc2F)c3cccs3 |
| IUPAC Name | methyl 2-(2,4-difluorophenyl)-5-thiophen-2-ylpyrazole-3-carboxylate |
| Molecular Weight (g/mol) | 320.314 |
| Formula Weight | 320.043 |