missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MethoCult™ H4535 Enriched Without EPO

Product Code. 17118231
Encompass
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 17118231

Brand: STEMCELL Technologies 04535

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methylcellulose-based medium with recombinant cytokines (without EPO) for human cells

  • MethoCult™ H4535 Enriched Without EPO (MethoCult™ GF+ H4535) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples
  • This medium supports optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and is recommended for use with CD34+ cells and other purified cell populations

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 100 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.