Learn More
Methanesulfonic Acid, TRC
Product Code. 30008451
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008451 250 mL 250mL
30007379 100 mL 100mL
30005510 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008451 Supplier Toronto Research Chemicals Supplier No. TRCM225940250ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-75-2
Molecular Formula C H4 O3 S
Molecular Weight (g/mol) 96.11
Synonym Methanesulfonic Acid, Lutropur MSA, Lutropure MAS, MCAT 1201, Mesylic acid, Methylsulfonic acid, NSC 3718, Scaleva
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O
Spezifikation
Quantity 250 mL
InChI Formula InChI=1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)
Formula Weight 95.9881
Percent Purity >95%
Chemical Name or Material Methanesulfonic Acid
Grade HPLC