Learn More
Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes
Shop All Nickel-Electro Products
Click to view available options
Length (Metric):
300 mm
340 mm
400 mm
430 mm
490 mm
Unit Size:
Each
Description
- Stainless steel square profile
- Includes lid
- Square shape prevents it from rolling off bench top
- Silicon in lid and base
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Sterility | Sterile |
| Quantity | Each |
| Length (Metric) | 340 mm |
| Type | Sterilizing Box |
| For Use With (Equipment) | Autoclave |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering