Learn More
Grant Instruments™ Metal Pump Connector, Dual Seal
Product Code. 15162595
Click to view available options
For Use With (Equipment):
Fits 4mm dia. Semi Rigid Tubing
Unit Size:
Each
Product Code. 15162595
Brand: Grant Instruments™ MSR4
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Fits semi-rigid tubing. Grant Instruments™ Metal Pump Connector is for use with Grant Refrigerated Baths and Heating Circulators
  • Metal pump inlet/outlet connector
  • Dual seal super rapid stlyle fits semi-rigid tubing
  • Temperature range -20 to 100°C
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fits 4mm dia. Semi Rigid Tubing
Temperature Range -20°C to +100°C
Type Pump Connector
Material Metal