Learn More
Lenz Laborglasinstrumente™ Metal Clamps
Click to view available options
Diameter (Metric):
60 mm
200 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1/Pk. |
| Type | Laboratory Clamp |
| For Use With (Equipment) | Fixing Lids Onto Reaction Vessels |
| Diameter (Metric) | 60 mm |