Learn More
meta-Methoxytopolin Riboside, TRC
Product Code. 30480133
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480133 250 mg 250mg
30473338 100 mg 100mg
30478599 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30480133 Supplier Toronto Research Chemicals Supplier No. TRCM271665250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101565-95-1
Molecular Formula C18H21N5O5
Molecular Weight (g/mol) 387.39
Synonym N-[(3-Methoxyphenyl)methyl]adenosine, MemTR, 6-(3-Methoxybenzylamino)-9-ß-dribofuranosylpurine, N-[(3-Methoxyphenyl)methyl]-adenosine
IUPAC Name (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[(3-methoxyphenyl)methylamino]purin-9-yl]oxolane-3,4-diol
SMILES COc1cccc(CNc2ncnc3c2ncn3[C@@H]4O[C@H](CO)[C@H](O)[C@@H]4O)c1
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C18H21N5O5/c1-27-11-4-2-3-10(5-11)6-19-16-13-17(21-8-20-16)23(9-22-13)18-15(26)14(25)12(7-24)28-18/h2-5,8-9,12,14-15,18,24-26H,6-7H2,1H3,(H,19,20,21)/t12-,14+,15+,18-/m1/s1
Formula Weight 387.1543
Chemical Name or Material meta-Methoxytopolin Riboside