Learn More
Mesoridazine Besylate, TRC
Product Code. 30480620
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480620 500 mg 500mg
30480278 250 mg 250mg
30470149 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30480620 Supplier Toronto Research Chemicals Supplier No. TRCM225755500MG
 Restricted Item
Estimated Shipment: 07-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32672-69-8
Molecular Formula C21 H26 N2 O S2 . C6 H6 O3 S
Molecular Weight (g/mol) 544.75
Synonym 10H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, benzenesulfonate (1:1)10H-Phenothiazine, 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-, monobenzenesulfonate (9CI), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine (1:1), Benzenesulfonic acid, compd. with 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylsulfinyl)phenothiazine (1:1) (8CI), Lidanar, Lidanil, Mesoridazine benzenesulfonate, Mesoridazine besylate, NC 123, Imp. B (EP) as Besilate: 10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulphinyl)-10H-phenothiazine Besilate (MesoridazineBesilate)
IUPAC Name benzenesulfonic acid;10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfinylphenothiazine
SMILES CN1CCCCC1CCN2c3ccccc3Sc4ccc(cc24)S(=O)C.OS(=O)(=O)c5ccccc5
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C21H26N2OS2.C6H6O3S/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(26(2)24)15-19(21)23;7-10(8,9)6-4-2-1-3-5-6/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3;1-5H,(H,7,8,9)
Formula Weight 544.1524
Chemical Name or Material Mesoridazine Besilate