Learn More
meso-Tetra (p-Bromophenyl) Porphine, TRC
Specifications
Specifications
| Chemical Name or Material | meso-Tetra (p-Bromophenyl) Porphine |
| CAS | 29162-73-0 |
| Molecular Formula | C44H26Br4N4 |
| Quantity | 250 mg |
| Synonym | 5,10,15,20-Tetrakis(4-bromophenyl)-21H,23H-porphine, 5,10,15,20-Tetra(4-bromophenyl)porphyrin, 5,10,15,20-Tetra(p-bromophenyl)porphyrin, 5,10,15,20-Tetrakis(4-bromophenyl)-21H,23H-porphyrin, 5,10,15,20-Tetrakis(4-bromophenyl)porphyrin, 5,10,15,20-Tetrakis(p-bromophenyl)porphyrin, Tetra(4-bromophenyl)porphyrin, Tetra(p-bromophenyl)porphyrin, Tetrakis(4-bromophenyl)porphyrin, meso-Tetra(4-bromophenyl)porphyrin, meso-Tetra(p-bromophenyl)porphyrin, meso-Tetrakis(4-bromophenyl)porphyrin |
| InChI Formula | InChI=1S/C44H26Br4N4/c45-29-9-1-25(2-10-29)41-33-17-19-35(49-33)42(26-3-11-30(46)12-4-26)37-21-23-39(51-37)44(28-7-15-32(48)16-8-28)40-24-22-38(52-40)43(36-20-18-34(41)50-36)27-5-13-31(47)14-6-27/h1-24,49,52H/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40- |
| SMILES | Brc1ccc(cc1)c2c3C=Cc([nH]3)c(c4ccc(Br)cc4)c5ccc(n5)c(c6ccc(Br)cc6)c7ccc([nH]7)c(c8ccc(Br)cc8)c9ccc2n9 |
| IUPAC Name | (5Z,10Z,14Z,19Z)-5,10,15,20-tetrakis(4-bromophenyl)-21,23-dihydroporphyrin |
| Molecular Weight (g/mol) | 930.32 |
| Formula Weight | 925.8891 |