missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MesenCult™-ACF Plus Medium Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- Reduce variability in your human mesenchymal stromal cell (MSC; also known as mesenchymal stem cell) cultures and improve experimental reproducibility by using this animal component-free (ACF) and extracellular vesicle (EV)-free medium
- MesenCult-ACF Plus Medium is optimized so you can derive human MSCs from multiple sources, such as bone marrow or adipose tissue, without serum
- Compared to serum-containing or EV-depleted serum-containing media, MSCs cultured with this kit expand more efficiently (rate and cumulative cell total) and without compromising function
- The cultured cells exhibit characteristic MSC surface marker expression and retain robust expansion rate and trilineage differentiation capacities
- This kit is part of a complete ACF workflow for deriving, expanding, and cryopreserving MSCs, as well as differentiating human pluripotent stem cells into mesenchymal progenitors that is optimized for efficient and consistent MSC cultures
Specifications
Specifications
| Cell Type | Mesenchymal Cells, PSC-Derived, Mesenchymal Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Expansion, Maintenance |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction