Learn More
(±)-Mepivacaine-d3 (N-methyl-d3), CDN
Product Code. 30319574
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319574
Brand: CDN CDND78610.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mepivacaine-d3
CAS 1346597-90-7
Molecular Formula C15 D3 H19 N2 O
Quantity 0.005 g
Synonym N-(2,6-Dimethylphenyl)-1-(trideuteriomethyl)piperidine-2-carboxamide, N-(2,6-Dimethylphenyl)-1-(methyl-d3)-2-piperidinecarboxamide, 1-(Methyl-d3)-2',6'-pipecoloxylidide, (±)-Mepivacaine-d3, Mepivacaine-d3, (±)-Mepivacaine-d3 (N-methyl-d3), (±)-Mepivacaine-d3 (N-methyl-d3)
InChI Formula InChI=1 S/C15H22N2O/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3/h6-8,13 H,4-5,9-10H2,1-3H3,(H,16,18)/i3D3
SMILES [2 H]C([2 H])([2 H])N1CCCCC1C(=O)Nc2c(C)cccc2C
IUPAC Name N-(2,6-dimethylphenyl)-1-(trideuteriomethyl)piperidine-2-carboxamide
Molecular Weight (g/mol) 249.367
Formula Weight 249.192 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity