Learn More
Epredia™ Menzel™ Microscope Coverslips
Product Code. 15707592 Shop All Epredia Products
Click to view available options
Length (Metric):
18 mm
20 mm
24 mm
Width (Metric):
18 mm
20 mm
24 mm
Unit Size:
Pack of 1000
Product Code. 15707592
Brand: Epredia™ BB02400240A153FST0
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Product Type Coverslip
Shape Square
Material Borosilicate Glass
Length (Metric) 24 mm
Width (Metric) 24 mm
Thickness (Metric) #1 (0.13 to 0.16 mm)
Dimensions (L x W) 24 x 24 mm
For Use With (Equipment) Automated equipment
Quantity 1000/Pk.