Learn More
Menthyl Anthranilate, TRC
Specifications
Specifications
| Chemical Name or Material | (1R,2S,5R) Menthyl Anthranilate |
| CAS | 307556-71-4 |
| Molecular Formula | C17 H25 N O2 |
| Quantity | 1 g |
| Synonym | (1R,2S,5R)-2-isopropyl-5-methylcyclohexyl 2-aminobenzoate |
| InChI Formula | InChI=1S/C17H25NO2/c1-11(2)13-9-8-12(3)10-16(13)20-17(19)14-6-4-5-7-15(14)18/h4-7,11-13,16H,8-10,18H2,1-3H3/t12-,13+,16-/m1/s1 |
| SMILES | [C@H](C)(C)[C@H]1[C@H](OC(=O)C2=C(N)C=CC=C2)C[C@H](C)CC1 |
| IUPAC Name | (1R,2S,5R)-2-isopropyl-5-methylcyclohexyl 2-aminobenzoate |
| Molecular Weight (g/mol) | 275.39 |
| Formula Weight | 275.19 |