missing translation for 'onlineSavingsMsg'
Learn More

Menthone, mixture of isomers, 98%

Product Code. 11413248
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11413248

Brand: Thermo Scientific Alfa Aesar A17665.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10458-14-7 | C10H18O | 154.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10458-14-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00062998
InChI Key NFLGAXVYCFJBMK-UHFFFAOYSA-N
Synonym isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone
PubChem CID 6986
ChEBI CHEBI:36742
IUPAC Name 5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC1CCC(C(=O)C1)C(C)C

Specifications

Density 0.894
Boiling Point ∼207°C
Flash Point 72°C (162°F)
Quantity 250 g
Formula Weight 154.25
Percent Purity 98%
Chemical Name or Material Menthone, mixture of isomers
Product Identifier
  • Menthone, mixture of isomers
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Menthone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.