missing translation for 'onlineSavingsMsg'
Learn More

Menthone Glycerin Acetal, TRC

Product Code. 30481475
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481475 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481475 Supplier Toronto Research Chemicals Supplier No. TRCM63187010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Menthone Glycerin Acetal
CAS 63187-91-7
Molecular Formula C13 H24 O3
Quantity 10 g
Synonym Menthone Glycerine Acetal, Menthone Glyceryl Ketal, [9-Methyl-6-(propan-2-yl)-1,4-dioxaspiro[4.5]decan-2-yl]methanol, 9-Methyl-6-(1-methylethyl)-1,4-dioxaspiro[4.5]decane-2-methanol, (6-Isopropyl-9-methyl-1,4-dioxaspiro[4.5]decan-2-yl)methanol, (9-Methyl-6-propan-2-yl-1,4-dioxaspiro[4.5]decan-3-yl)methanol, 6-Isopropyl-9-methyl-1,4-dioxaspiro[4,5]decane-2-methanol, Fema Gras 3808, Frescolat MGA
InChI Formula InChI=1S/C13H24O3/c1-9(2)12-5-4-10(3)6-13(12)15-8-11(7-14)16-13/h9-12,14H,4-8H2,1-3H3
SMILES CC(C)C1CCC(C)CC12OCC(CO)O2
IUPAC Name (9-methyl-6-propan-2-yl-1,4-dioxaspiro[4.5]decan-3-yl)methanol
Molecular Weight (g/mol) 228.328
Formula Weight 228.173
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.