Learn More
STEMCELL Technologies MEM with Earle's & NEAA
Description
- Minimum Essential Medium Eagle with Earle's salts and non-essential amino acids (NEAA) is recommended for a wide variety of cell culture applications
- Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 500 mL |